missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF500 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58239
This item is not returnable.
View return policy
Description
ZNF500 Polyclonal specifically detects ZNF500 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF500 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26048 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ZNF500 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EEPRCMDPAQRDAPLENEGPGIQLEDGGDGREDAPLRMEWYRVLSARCQGPGHPLPGQRPAPVRGLVRPDQPRG | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction