missing translation for 'onlineSavingsMsg'
Learn More

ZNF50 Polyclonal Antibody

ZNF50 Polyclonal Antibody

Brand:  Invitrogen PA537087

Product Code. 15544241

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF50 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF50
Polyclonal
Unconjugated
ZSCAN22
AW227621; D530006B18Rik; GLI-Kruppel family member HKR2; HKR2; Krueppel-related zinc finger protein 2; LOC102553002; Protein HKR2; zinc finger and SCAN domain containing 22; zinc finger and SCAN domain-containing protein 22; Zinc finger protein 50; zinc finger protein 8-like; ZNF50; ZSCAN22
Rabbit
Antigen Affinity Chromatography
RUO
102553002, 232878, 342945
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
P10073
ZSCAN22
Synthetic peptide corresponding to amino acids 363-408 of human ZNF50.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.