missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF497 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569060
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
May be involved in transcriptional regulation.
Specifications
| ZNF497 | |
| Polyclonal | |
| Unconjugated | |
| ZNF497 | |
| Zinc finger protein 497; zinc finger-like protein; ZNF497 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 162968 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ZNH5 | |
| ZNF497 | |
| synthetic peptide directed towards the N terminal region of human ZNF497. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction