missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF483 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-31592-25ul
This item is not returnable.
View return policy
Description
ZNF483 Polyclonal specifically detects ZNF483 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZNF483 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q8TF39 | |
| ZNF483 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LLENLRNLEFLDFPVSKLELISQLKWVELPWLLEEVSKSSRLDESALDKIIERCLRDDDHGLMEESQQYCGSSEEDHGNQGNSKG | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BMZF-2zinc finger 2, bone marrow, BMZF2zinc finger protein 27 (KOX 22), Bone marrow zinc finger 2, KOX22Zinc finger protein 233, zinc finger protein 224, Zinc finger protein 255Zinc finger protein KOX22, Zinc finger protein 27, ZNF255zinc finger protein ZNF255, ZNF27ZNF233 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 158399 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction