missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF48 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554891
This item is not returnable.
View return policy
Description
Immunogen sequence: GLGSENVISQ PNEFEHTPQE DDLGFKEEDL APDHEVGNVS LKPEGIQNWD DLWVQREGLG KPQPRDRGPR LLGEPRWGQA SSDRA Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 68%.
ZNF48, also named as zinc finger protein 48, is 618 amino acid protein, which contains twelves C2H2-type zinc fingers and belongs to the Krueppel C2H2-type zinc-finger protein family. ZNF48 localizes in the nucleus and may be involves in transcription regulation.
Specifications
| ZNF48 | |
| Polyclonal | |
| Unconjugated | |
| ZNF48 | |
| Zinc finger protein 48; zinc finger protein 553; ZNF48; ZNF553 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 197407 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MX3 | |
| ZNF48 | |
| Recombinant protein corresponding to Human ZNF48. Recombinant protein control fragment (Product #RP-93626). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction