Learn More
Invitrogen™ ZNF471 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595797
Description
Immunogen sequence: TRSPFSDWES IYVTQELPLK QFMYDDACME GITSYGLECS TFEENWKWED LFEKQMGSHE MFSKKEIITH KETITKETEF KYTKFGKCIH LENIEESIYN HTSDKKSFSK NSMVIKHKKV YVGKKLFKCN E; Positive Samples: Mouse pancreas, Mouse skeletal muscle, Mouse brain, Rat pancreas, Rat skeletal muscle, Rat brain; Cellular Location: Nucleus.
ZNF471 (Zinc Finger Protein 471) is a Protein Coding gene. Among its related pathways are Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and DNA-binding transcription factor activity.
Specifications
| ZNF471 | |
| Polyclonal | |
| Unconjugated | |
| ERP1; EZFIT-related protein 1; KIAA1396; Z1971; zinc finger protein 471; ZNF471 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-210 of human ZNF471 (NP_0658642). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.