missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF462 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567676
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
May be involved in transcriptional regulation.
Specifications
| ZNF462 | |
| Polyclonal | |
| Unconjugated | |
| ZNF462 | |
| 6030417H05; 9430078C22Rik; DKFZP762N2316; gene trap insertion site 4-2; Gt4-2; KIAA1803; RGD1562123; Zfp462; ZFPIP; Zinc finger PBX1-interacting protein; Zinc finger protein 462; ZNF462 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 242466, 362522, 58499 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| Q96JM2 | |
| Zfp462, ZNF462 | |
| Synthesized peptide derived from human ZNF462. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction