missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF451 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569000
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 87%; Rat: 100%.
E3 SUMO-protein ligase; has a preference for SUMO2 and SUMO3 and facilitates UBE2I/UBC9-mediated sumoylation of target proteins (PubMed:26524493, PubMed:26524494). Plays a role in protein SUMO2 modification in response to stress caused by DNA damage and by proteasome inhibitors (in vitro). Required for MCM4 sumoylation. Has no activity with SUMO1 (PubMed:26524493). Preferentially transfers an additional SUMO2 chain onto the SUMO2 consensus site 'Lys-11' (PubMed:26524493). Negatively regulates transcriptional activation mediated by the SMAD4 complex in response to TGF-beta signaling. Inhibits EP300-mediated acetylation of histone H3 at 'Lys-9' (PubMed:24324267). Plays a role in regulating the transcription of AR targets (PubMed:18656483). [UniProt].
Specifications
| ZNF451 | |
| Polyclonal | |
| Unconjugated | |
| ZNF451 | |
| 4930515K21Rik; 4933435G09Rik; AI596398; Coactivator for steroid receptors; COASTER; dJ417I1.1; E3 SUMO-protein ligase ZNF451; E3 SUMO-protein ligase ZNF451; zinc finger protein 451; E3 SUMO-protein transferase ZNF451; KIAA0576; Kiaa0576-hp; KIAA1702; mKIAA1702; Zfp451; Zinc finger protein 451; Zinc finger protein 451-like protein; Znf451 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26036 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y4E5 | |
| ZNF451 | |
| synthetic peptide directed towards the C terminal region of human ZNF451. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido