missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF449 Antibody (OTI4C6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74952CL1
This item is not returnable.
View return policy
Description
ZNF449 Monoclonal antibody specifically detects ZNF449 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| ZNF449 | |
| Monoclonal | |
| CoraFluor 1 | |
| HZF1Zinc finger protein HZF1, KIAA2007, zinc finger protein 1, zinc finger protein 266 | |
| Full length human recombinant protein of human ZNF449 (NP_689908) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI4C6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 203523 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction