missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF447 Monoclonal Antibody (OTI2A9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526114
This item is not returnable.
View return policy
Description
ZNF447 Monoclonal Antibody for Western Blot, IHC (P)
May be involved in transcriptional regulation.
Specifications
| ZNF447 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TBC5 | |
| ZSCAN18 | |
| Full length human recombinant protein of ZSCAN18 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A9 | |
| Unconjugated | |
| ZSCAN18 | |
| EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65982 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction