Learn More
Invitrogen™ ZNF447 Monoclonal Antibody (2H1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522540
Description
Peptide Sequence: MLPLEKAFAS PRSSPAPPDL PTPGSAAGVQ QEEPETIPER TPADLEFSRL RFREFVYQEA AGPHQTLARL HELCRQWLMP EARSKEQMLE LLVLEQFLGI LPDKVRPWVV AQYPESCKKA ASLVEGLADV LEEPGMLLGS PAGSSSILSD GVYERHMDPL LLPGELASPS QALGAGEIPA PSETPWLSPD PLFLEQRRVR EAKTEEDGPA NTEQKLKSFP EDPQHLGEWG HLDPAEENLK SYRKLLLWGY QLSQPDAASR LDTEELRLVE RDPQGSSLPE GGRRQESAGC ACEEAAPAGV LPELPTEAPP GDALADPPSG TTEEEEEQPG KAPDPQDPQD AESDSATGSQ RQSVIQQPAP DRGTAKLGTK RPHPEDGDEQ SLEGVSSSGD SAGLEAGQGP GADEPGLSRG KPYACGECGE AFAWLSHLME HHSSHGGRKR YACQGCWKTF HFSLALAEHQ KTHEKEKSYA LGGARGPQPS TREAQAGARA GGPPESVEGE APPAPPEAQR.
May be involved in transcriptional regulation.
Specifications
| ZNF447 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8TBC5 | |
| ZSCAN18 | |
| ZSCAN18 (AAH22846, 1 a.a. ~ 510 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 2H1 | |
| Unconjugated | |
| ZSCAN18 | |
| EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 65982 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.