missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF445 Polyclonal specifically detects ZNF445 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ZNF445 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | zinc finger protein 168, zinc finger protein 445, zinc finger protein with KRAB and SCAN domains 15, ZKSCAN15, ZNF168 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_775540). Peptide sequence KLHHKEVYKQEKRQEDFKQSYRQSQVISTVEKTFPCQNCGKTFTQKKSLI |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?