missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF444 Polyclonal antibody specifically detects ZNF444 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | ZNF444 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunoprecipitation |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HZF3, MGC126498, zinc finger protein 268, zinc finger protein 3, Zinc finger protein HZF3 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 200 and 250 of human Zinc Finger Protein 444 using the numbering given in entry NP_060807.2 (GeneID 55311). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?