missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF415 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98201G
This item is not returnable.
View return policy
Description
ZNF415 Polyclonal antibody specifically detects ZNF415 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ZNF415 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| MGC126536, MGC141923, zinc finger protein 287, ZKSCAN13Zinc finger protein with KRAB and SCAN domains 13 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF415 fragment. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 55786 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction