missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals ZNF41 Antibody (1E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00007592-M05
This item is not returnable.
View return policy
Description
ZNF41 Monoclonal antibody specifically detects ZNF41 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ZNF41 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| zinc finger protein 296, Zinc finger protein 342ZFP296ZNF342 | |
| ZNF41 (NP_009061, 221 a.a. ~ 321 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GNNFPHSPSSTKNENAKTGANSCEHDHYEKHLSHKQAPTHHQKIHPEEKLYVCTECVMGFTQKSHLFEHQRIHAGEKSRECDKSNKVFPQKPQVDVHPSV* | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1E1 | |
| Western Blot, ELISA | |
| NP_009061 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 7592 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction