missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF408 Mouse anti-Human, Clone: PCRP-ZNF408-1E5, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | ZNF408 |
|---|---|
| Clone | PCRP-ZNF408-1E5 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array |
| Applications | Western Blot, Flow Cytometry, Peptide Array |
Description
ZNF408 Monoclonal specifically detects ZNF408 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.Specifications
| ZNF408 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25 | |
| Recombinant full-length human ZNF408 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-ZNF408-1E5 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 79797 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title