missing translation for 'onlineSavingsMsg'
Learn More

ZNF408 Mouse anti-Human, Clone: PCRP-ZNF408-1E5, Novus Biologicals™

Product Code. 18350662 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350662 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350662 Supplier Novus Biologicals Supplier No. NBP30736620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF408 Monoclonal specifically detects ZNF408 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF408
Applications Western Blot, Flow Cytometry, Peptide Array
Classification Monoclonal
Clone PCRP-ZNF408-1E5
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25
Host Species Mouse
Immunogen Recombinant full-length human ZNF408 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79797
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.