missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF404 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584605
This item is not returnable.
View return policy
Description
Immunogen sequence: VSLDFNFTTE SNKLSSEKRN YEVNAYHQET WKRNKTFNLM RFIFRTDPQY TI Highest antigen sequence indentity to the following orthologs: Mouse - 29%, Rat - 28%.
ZNF404 (Zinc Finger Protein 404) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
Specifications
| ZNF404 | |
| Polyclonal | |
| Unconjugated | |
| ZNF404 | |
| Zinc finger protein 404; ZNF404 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 342908 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q494X3 | |
| ZNF404 | |
| Recombinant protein corresponding to Human ZNF404. Recombinant protein control fragment (Product #RP-108491). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction