Learn More
ZNF397 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA538738
Product Code. 15565651
  • £416.00 / 100µg
Description
This gene encodes a protein with a N-terminal SCAN domain, and the longer isoform contains nine C2H2-type zinc finger repeats in the C-terminal domain. The protein localizes to centromeres during interphase and early prophase, and different isoforms can repress or activate transcription in transfection studies. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF397
Polyclonal
Unconjugated
ZNF397
ZNF47, ZSCAN15, zinc finger and SCAN domain-containing protein 15, zinc finger protein 47
Rabbit
Antigen affinity chromatography
RUO
84307
-20°C
Liquid
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NF99
ZNF397
A synthetic peptide derived from the N-terminal region of human ZNF397