missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF396 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582258
This item is not returnable.
View return policy
Description
Immunogen sequence: FEKRQGNPSW KKQQKCDECG KIFSQSSALI LHQRIHSGKK PYACDECAKA FSRSAILIQH RRTHTDSKYE HAIAEAQKNM YFKTRLKCPR SLSG Highest antigen sequence indentity to the following orthologs: Mouse - 51%, Rat - 51%.
Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
Specifications
| ZNF396 | |
| Polyclonal | |
| Unconjugated | |
| ZNF396 | |
| Zinc finger and SCAN domain-containing protein 14; zinc finger protein 396; ZNF396; ZSCAN14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 252884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96N95 | |
| ZNF396 | |
| Recombinant protein corresponding to Human ZNF396. Recombinant protein control fragment (Product #RP-89422). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction