missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF385D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557566
This item is not returnable.
View return policy
Description
Immunogen sequence: HAKKLKALEA MKNKQKSVTA KDSAKTTFTS ITTNTINTSS DKTDGTAGTP AISTTTTVEI RKSSVMTTEI TSKVE Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 73%.
ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.
Specifications
| ZNF385D | |
| Polyclonal | |
| Unconjugated | |
| ZNF385D | |
| si:dkey-82o10.2; Zfp385d; Zinc finger protein 385D; zinc finger protein 659; ZNF385D; ZNF659 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H6B1 | |
| ZNF385D | |
| Recombinant protein corresponding to Human ZNF385D. Recombinant protein control fragment (Product #RP-96376). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction