missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF385D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79397
This item is not returnable.
View return policy
Description
ZNF385D Polyclonal specifically detects ZNF385D in Human samples. It is validated for Western Blot.
Specifications
| ZNF385D | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ12586, zinc finger protein 329 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79750 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_078973 | |
| ZNF385D | |
| Synthetic peptide directed towards the N terminal of human ZNF385DThe immunogen for this antibody is ZNF385D. Peptide sequence ALEAMKNKQKSVTAKDSAKTTFTSITTNTINTSSDKTDGTAGTPAISTTT. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 92%; Equine: 92%; Pig: 92%; Rabbit: 92%; Chicken: 85%. | |
| Human, Rat, Pig, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction