missing translation for 'onlineSavingsMsg'
Learn More

ZNF385B Antibody (OTI1H10), DyLight 594, Novus Biologicals™

Product Code. 18409297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409297 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409297 Supplier Novus Biologicals Supplier No. NBP274948DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF385B Monoclonal antibody specifically detects ZNF385B in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF385B
Applications Western Blot
Classification Monoclonal
Clone OTI1H10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias C2H2-like zinc finger protein rearranged in thyroid adenomas, DKFZp686L0787, KRAB zinc finger protein, zinc finger protein 331, Zinc finger protein 463, ZNF463RITAZNF361Zinc finger protein 361
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 149-399 of human ZNF385B (NP_689733) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151126
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.