missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF385B Antibody (OTI1H10), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74948AF350
This item is not returnable.
View return policy
Description
ZNF385B Monoclonal specifically detects ZNF385B in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ZNF385B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ZNF385B | |
| Recombinant protein fragment corresponding to amino acids 149-399 of human ZNF385B (NP_689733) produced in E.coli. | |
| 0.1 mL | |
| 151126.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1H10 | |
| Western Blot | |
| C2H2-like zinc finger protein rearranged in thyroid adenomas, DKFZp686L0787, KRAB zinc finger protein, zinc finger protein 331, Zinc finger protein 463, ZNF463RITAZNF361Zinc finger protein 361 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction