missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF385A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540558
This item is not returnable.
View return policy
Description
Peptide sequence: EPAPTLGLFS NYSTMDPVQK AVLSHTFGGP LLKTKRPVIS CNICQIRFNS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
ZNF385 is a new candidate transcription factor.
Specifications
| ZNF385A | |
| Polyclonal | |
| Unconjugated | |
| ZNF385A | |
| G-protein coupled receptor 84; zinc finger protein 385A; hematopoietic zinc finger protein; Hzf; LOW QUALITY PROTEIN: zinc finger protein 385A; retinal zinc finger protein; RZF; si:dkey-228g21.6; wu:fj44b04; ZFP385; Zfp385a; zinc finger protein 385; zinc finger protein 385A; ZNF385; Znf385a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25946 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96PM9 | |
| ZNF385A | |
| Synthetic peptide directed towards the N-terminal of human ZNF385 (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction