Learn More
Invitrogen™ ZNF38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590386
Description
Immunogen sequence: PGHQVSTPPN EQKPVWEKIS SSGTAKESPS SMQPQPLETS HKYESWGPLY IQESGEEQEF AQDPRKVRDC RLSTQHEESA DEQKGSEAEG LKGDIISVII ANKPEASLER QCVNLENEKG TKPPLQEAGS KKGRESVPTK PTPGERRYIC A; Positive Samples: MCF7, 293T, Mouse brain, Mouse heart, Rat brain, Rat heart; Cellular Location: Nucleus.
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
Specifications
| ZNF38 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN21 | |
| AI326272; CTfin51; hypothetical protein LOC527103; NY-REN-21; renal carcinoma antigen NY-REN-21; RU49; Transcription factor RU49; Zfp38; zfp-38; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 3; zinc finger protein with KRAB and SCAN domains 1; zinc finger proliferation 1; zinc finger protein 38; zinc finger protein 38 (KOX 25); zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen; zinc finger with KRAB and SCAN domains 1; Zipro1; ZKSCAN1; ZNF38; ZSCAN21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22697, 304342, 7589 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q07231, Q9Y5A6 | |
| ZSCAN21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-280 of human ZSCAN21 (NP_6660191). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.