missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZNF37A Antibody (4H9), Novus Biologicals™

Product Code. 18343959 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343959 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343959 Supplier Novus Biologicals™ Supplier No. H00007587M03100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF37A Monoclonal antibody specifically detects ZNF37A in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF37A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4H9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001007095
Gene Alias KOX32zinc finger protein 34 (KOX 32), MGC10520, zinc finger protein 34, Zinc finger protein KOX32
Host Species Mouse
Immunogen ZNF37A (NP_001007095, 121 a.a. ∽ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NKNGNSFWLNEDLIWHQKIKNWEQSFEYNECGKAFPENSLFLVHKRGYTGQKTCKYTEHGKTCDMSFFITHQQTHPRENHYGNECGENIFEESILLEHQSVYPFSQKLNL
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7587
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.