missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF367 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564961
This item is not returnable.
View return policy
Description
Immunogen sequence: RHCPKHPYARL KREEPTDTLS KHQAADNKAA AEWLARYWEM REQRTPTLKG KLVQKADQEQ QDPLEYLQSD EEDDEKRGAQ RRLQEQRERL HGALALIELA NLTG Highest antigen sequence identity to the following orthologs - mouse 95%, rat 96%.
Transcriptional activator. Isoform 1 may be involved in transcriptional activation of erythroid genes.
Specifications
| ZNF367 | |
| Polyclonal | |
| Unconjugated | |
| ZNF367 | |
| 8030486J21Rik; AFF29; C2H2 zinc finger protein ZFF29; C2H2 zinc finger protein ZFF29a; CDC14 cell division cycle 14 homolog B; CDC14B; habp4; hyaluronan binding protein 4; hypothetical protein LOC550421; LOW QUALITY PROTEIN: zinc finger protein 367; ZFF29; ZFF29a; ZFF29b; Zfp367; zgc:112157; Zinc finger protein 367; Znf367 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 195828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7RTV3 | |
| ZNF367 | |
| Recombinant Human ZNF367. Recombinant protein control fragment (Product #RP-91324). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction