missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF366 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5110886
This item is not returnable.
View return policy
Description
Immunogen sequence: STKSEHAPEV LEEACKEEKE DASKGEWEKR SKGDLGAEGG QERDCAGRDE CLSLRAFQST RRGPSFSDYL YFKHRDESLK ELLERK
Has transcriptional repression activity. Acts as corepressor of ESR1; the function seems to involve CTBP1 and histone deacetylases. [UniProt]
Specifications
| ZNF366 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N895 | |
| Znf366 | |
| Recombinant Protein corresonding to Human ZNF366 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| Znf366 | |
| DCSCRIPT; DC-SCRIPT; Dendritic cell-specific transcript protein; Zinc finger protein 366; ZNF366 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction