missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF115 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110303
This item is not returnable.
View return policy
Description
Immunogen sequence: SPKLPEYICP RCESGFIEEV TDDSSFLGGG GSRIDNTTTT HFAELWGHLD HTMFFQDFRP FLSSSPLDQD NRANERGHQT HTDFWGARPP RLPLGRRYRS RGSSRPDRSP A.
ZNF364 contains 1 RING-type zinc finger. The exact function of ZNF364 remains unknown.
Specifications
| RNF115 | |
| Polyclonal | |
| Unconjugated | |
| RNF115 | |
| 2610028E05Rik; AU042696; BCA2; CL469780; E3 ubiquitin-protein ligase RNF115; Rab7-interacting RING finger protein; rabring 7; Rabring7; ring finger protein 115; RING-type E3 ubiquitin transferase RNF115; RNF115; Zfp364; Zinc finger protein 364; ZNF364 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 362002 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| 0 | |
| RNF115 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human RNF115 (NP_055270.1). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction