missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZNF364 Antibody, Novus Biologicals™

Product Code. 18346379 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346379 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346379 Supplier Novus Biologicals™ Supplier No. H00027246B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ZNF364 Polyclonal antibody specifically detects ZNF364 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF364
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_055270.1
Gene Alias E3 ubiquitin-protein ligase RNF115, EC 6.3.2.-, Rabring 7, ring finger protein 115CL469780, Zinc finger protein 364BCA2, ZNF364
Host Species Mouse
Immunogen RNF115 (NP_055270.1, 1 a.a. - 304 a.a.) full-length human protein. MAEASAAGADSGAAVAAHRFFCHFCKGEVSPKLPEYICPRCESGFIEEVTDDSSFLGGGGSRIDNTTTTHFAELWGHLDHTMFFQDFRPFLSSSPLDQDNRANERGHQTHTDFWGARPPRLPLGRRYRSRGSSRPDRSPAIEGILQHIFAGFFANSAIPGSPHPFSWSGMLHSNPGDYAWGQTGLDAIVTQLLGQLENTGPPPADKEKITSLPTVTVTQEQVDMGLECPVCKEDYTVEEEVRQLPCNHFFHSSCIVPWLELHDTCPVCRKSLNGEDSTRQSQSTEASASNRFSNDSQLHDRWTF
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 27246
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.