missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF364 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94360-0.02ml
This item is not returnable.
View return policy
Description
ZNF364 Polyclonal antibody specifically detects ZNF364 in Rat samples. It is validated for Western Blot
Specifications
| ZNF364 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| E3 ubiquitin-protein ligase RNF115, EC 6.3.2.-, Rabring 7, ring finger protein 115CL469780, Zinc finger protein 364BCA2, ZNF364 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human RNF115 (NP_055270.1). SPKLPEYICPRCESGFIEEVTDDSSFLGGGGSRIDNTTTTHFAELWGHLDHTMFFQDFRPFLSSSPLDQDNRANERGHQTHTDFWGARPPRLPLGRRYRSRGSSRPDRSPA | |
| 0.02 mL | |
| Breast Cancer, Cancer, Zinc Finger | |
| 27246 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction