missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF354C Antibody (Macrogenics patent anti-KID3) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28380JF635
This item is not returnable.
View return policy
Description
ZNF354C Monoclonal antibody specifically detects ZNF354C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ZNF354C | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FLJ10390, ZFEND, zinc finger protein 358 | |
| KID3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Macrogenics patent anti-KID3 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 30832 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction