missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF350 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581951
This item is not returnable.
View return policy
Description
Immunogen sequence: FKLEQGEQLW TIEDGIHSGA CSDIWKVDHV LERLQSESLV NRRKPCHEHD AFENIVHCSK SQFLLGQNHD IFDLRGKSLK SNLTLVNQSK GYEIKNSVEF TGNGDSFLHA NHERLHTAIK FPASQ Highest antigen sequence indentity to the following orthologs: Mouse - 25%, Rat - 25%.
ZBRK1 is a member of the Kruppel-associated box-zinc finger protein (KRAB-ZFP) protein family of transcriptional repressors. ZBRK1 has been shown to not only be up-regulated in senescent fibroblasts but also interacts with BRCA1, a transcriptional regulator with tumor suppressor activity. ZBRK1 binds to a specific sequence within the GADD45 gene, supporting the assembly of a nuclear complex containing BRCA1 and repressing its transcription. Recent experiments demonstrate that ZBRK1 can homo-oligomerize in vitro and in vivo through a C-terminal transcriptional repression domain (CTRD), and this oligomerization facilitates the ZBRK1-directed transcriptional repression through ZBRK1 response elements. These results suggest that higher order ZBRK1 oligomers may assemble through both protein-DNA and CTRD-dependent protein-protein interactions.
Specifications
| ZNF350 | |
| Polyclonal | |
| Unconjugated | |
| ZNF350 | |
| KRAB zinc finger protein ZFQR; ZBRK1; ZFQR; Zinc finger and BRCA1-interacting protein with a KRAB domain 1; zinc finger protein 350; Zinc finger protein ZBRK1; zinc-finger protein ZBRK1; ZNF350 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 59348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9GZX5 | |
| ZNF350 | |
| Recombinant protein corresponding to Human ZNF350. Recombinant protein control fragment (Product #RP-99731). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction