missing translation for 'onlineSavingsMsg'
Learn More

ZNF350 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18414106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414106 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414106 Supplier Novus Biologicals Supplier No. NBP176344DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF350 Polyclonal antibody specifically detects ZNF350 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF350
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Berg36, BRF1zinc finger protein 36, C3H1 type-like 1, Butyrate response factor 1, cMG1, ERF1ERF-1, Protein TIS11B, RNF162BEGF-response factor 1, TIS11Bearly response factor Berg36, zinc finger protein 36, C3H type-like 1, zinc finger protein, C3H type, 36-like 1
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of the human ZBRK1. The immunogen is located within amino acids 410 - 460 of ZBRK1. Amino Acid Squence: KRIHTREKQEAKKVENPPAE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 59348
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.