missing translation for 'onlineSavingsMsg'
Learn More

ZNF350 Antibody (1A9), Novus Biologicals™

Product Code. 18407628 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407628 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407628 Supplier Novus Biologicals Supplier No. H00059348M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF350 Monoclonal antibody specifically detects ZNF350 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF350
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1A9
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH09921
Gene Alias Berg36, BRF1zinc finger protein 36, C3H1 type-like 1, Butyrate response factor 1, cMG1, ERF1ERF-1, Protein TIS11B, RNF162BEGF-response factor 1, TIS11Bearly response factor Berg36, zinc finger protein 36, C3H type-like 1, zinc finger protein, C3H type, 36-like 1
Host Species Mouse
Immunogen ZNF350 (AAH09921, 1 a.a. ∽ 532 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MIQAQESITLEDVAVDFTWEEWQLLGAAQKDLYRDVMLENYSNLVAVGYQASKPDALFKLEQGEQLWTIEDGIHSGACSDIWKVDHVLERLQSESLVNRRKPCHEHDAFENIVHCSKSQFLLGQNHDIFDLRGKSLKSNLTLVNQSKGYEIKNSVEFTGNGDSFLHANHERLHTAIKFPASQKLISTKSQFISPKHQKTRKLEKHHVCSECGKAFIKKSWLTDHQVMHTGEKPHRCSLCEKAFSRKFMLTEHQRTHTGEKPYECPECGKAFLKKSRLNIHQKTHTGEKPYICSECGKGFIQKGNLIVHQRIHTGEKPYICNECGKGFIQKTCLIAHQRFHTGKTPFVCSECGKSCSQKSGLIKHQRIHTGEKPFECSECGKAFSTKQKLIVHQRTHTGERPYGCNECGKAFAYMSCLVKHKRIHTREKQEAAKVENPPAERHSSLHTSDVMQEKNSANGATTQVPSVAPQTSLNISGLLANRNVVLVGQPVVRCAASGDNRGFAQDRNLVNAVNVVVPSVINYVLFYVTENP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 59348
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.