missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF35 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16935-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF35 is a new candidate transcription factor.Specifications
| ZNF35 | |
| Polyclonal | |
| Unconjugated | |
| ZNF35 | |
| HF.10, HF10, Zfp105, zinc finger protein 35, Zinc finger protein HF.10, ZNF35 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22694, 307547, 7584 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13682 | |
| Zfp35, ZNF35 | |
| ZNF35 Fusion Protein Ag10432 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction