missing translation for 'onlineSavingsMsg'
Learn More

ZNF35 Antibody (2B4), Novus Biologicals™

Product Code. 18351829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351829 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351829 Supplier Novus Biologicals Supplier No. H00007584M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF35 Monoclonal antibody specifically detects ZNF35 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF35
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2B4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13597
Gene Alias BRF2, Butyrate response factor 2, ERF2ERF-2, Protein TIS11D, RNF162Czinc finger protein 36, C3H1 type-like 2, TIS11DEGF-response factor 2, zinc finger protein 36, C3H type-like 1, zinc finger protein 36, C3H type-like 2, zinc finger protein, C3H type, 36-like 2
Host Species Mouse
Immunogen ZNF35 (AAH13597, 1 a.a. ~ 519 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MALAPWGPVKVKKEEEEEENFPGQASSQQVHSENIKVWAPVQGLQTGLDGSEEEEKGQNISWDMAVVLKATQEAPAASTLGSYSLPGTLAKSEILETHGTMNFLGAETKNLQLLVPKTEICEEAEKPLIISERIQKADPQGPELGEACEKGNMLKRQRIKREKKDFRQVIVNDCHLPESFKEEENQKCKKSGGKYSLNSGAVKNPKTQLGQKPFTCSVCGKGFSQSANLVVHQRIHTGEKPFECHECGKAFIQSANLVVHQRIHTGQKPYVCSKCGKAFTQSSNLTVHQKIHSLEKTFKCNECEKAFSYSSQLARHQKVHITEKCYECNECGKTFTRSSNLIVHQRIHTGEKPFACNDCGKAFAQSANLIVHQRSHTGEKPYECKECGKAFSCFSHLIVHQRIHTAEKPYDCSECGKAFSQLSCLIVHQRIHSGDLPYVCNECGKAFTCSSYLLIHQRIHNGEKPYTCNECGKAFRQRSSLTVHQRTHTGEKPYECEKCGAAFISNSHLMRHHRTHLVE
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7584
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.