missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF346 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582630
This item is not returnable.
View return policy
Description
Immunogen sequence: LMARYGRLAD PAVTDFPAGK GYPCKTCKIV LNSIEQYQAH VSGFKHKNQS PKTVASSLGQ IPMQRQPIQK DSTTL Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 79%.
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.
Specifications
| ZNF346 | |
| Polyclonal | |
| Unconjugated | |
| ZNF346 | |
| double-stranded RNA-binding zinc finger protein JAZ; Jaz; Just another zinc finger protein; Zfp346; Zinc finger protein 346; ZNF346 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23567 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UL40 | |
| ZNF346 | |
| Recombinant protein corresponding to Human ZNF346. Recombinant protein control fragment (Product #RP-92029). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction