missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF346 Polyclonal Antibody

Product Code. 12809192
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809192 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809192 Supplier Invitrogen™ Supplier No. PA521041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human kidney tissue lysate. PA5-21041 can be used with blocking peptide PEP-1155.

Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF346
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZNF346
Gene Accession No. Q9R0B7, Q9UL40
Gene Alias double-stranded RNA-binding zinc finger protein JAZ; Jaz; Just another zinc finger protein; Zfp346; Zinc finger protein 346; ZNF346
Gene Symbols Zfp346, ZNF346
Host Species Rabbit
Immunogen A 20 amino acid peptide near the amino terminus of human ZNF346.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23567, 26919, 306765
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.