missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF345 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20996
This item is not returnable.
View return policy
Description
ZNF345 Polyclonal specifically detects ZNF345 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZNF345 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| FLJ38524, Zfp-37, zinc finger protein 37 homolog, zinc finger protein 37 homolog (mouse), zinc finger protein homologous to Zfp37 in mouse, Zinc finger protein-37, mouse, homolog of, ZNF906 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Monkey (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q14585 | |
| ZNF345 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF345. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 25850 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction