missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563892
This item is not returnable.
View return policy
Description
Immunogen sequence: QETFGEEDPQ GSEPVEACDH ISKSEGSLEK LVEQRGPRAV TLTNGESSRE SGGNLRLL Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 31%.
May be involved in transcriptional regulation.
Specifications
| ZNF34 | |
| Polyclonal | |
| Unconjugated | |
| ZNF34 | |
| KOX32; Zinc finger protein 34; zinc finger protein 34 (KOX 32); zinc finger protein KOX32; ZNF34 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80778 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZ26 | |
| ZNF34 | |
| Recombinant protein fragment of Human ZNF34 (Product #RP-101591) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction