missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF33A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536013
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF33A belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF33A may be involved in transcriptional regulation.
Specifications
| ZNF33A | |
| Polyclonal | |
| Unconjugated | |
| ZNF33A | |
| brain my041 protein; KIAA0065; KOX2; KOX31; KOX5; NF11A; Zinc finger and ZAK-associated protein with KRAB domain; zinc finger protein 11A; Zinc finger protein 33A; zinc finger protein 33a (KOX 31); zinc finger protein KOX31; ZNF11; ZNF11A; ZNF33; ZNF33A; ZZAPK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q06730 | |
| ZNF33A | |
| Recombinant protein encompassing a sequence within the center region of human ZNF33A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction