missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF335 Polyclonal Antibody

Description
Immunogen sequence: RNGHLKFHIQR LHSPDGRKSG TPTARAPTQT PTQTIILNSD DETLATLHTA LQSSHGVLGP ERLQQALSQE HIIVAQEQ Highest antigen sequence identity to the following orthologs - mouse 85%, rat 82%.
Component or associated component of some histone methyltransferase complexes may regulate transcription through recruitment of those complexes on gene promoters. Enhances ligand-dependent transcriptional activation by nuclear hormone receptors. Plays an important role in neural progenitor cell proliferation and self-renewal through the regulation of specific genes involved brain development, including REST. Also controls the expression of genes involved in somatic development and regulates, for instance, lymphoblast proliferation.
Specifications
Specifications
| Antigen | ZNF335 |
| Applications | ChIP Assay, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF335 |
| Gene Accession No. | Q9H4Z2 |
| Gene Alias | 1810045J01Rik; LOW QUALITY PROTEIN: zinc finger protein 335; MCPH10; Nif; NIF1; NIF-1; NIF2; NRC-interacting factor 1; Zfp335; Zinc finger protein 335; zinc-finger/leucine-zipper co-transducer NIF1; zinc-finger/leucine-zipper co-transducer NIF-1; Znf335 |
| Gene Symbols | ZNF335 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?