missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF334 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-13567
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
ZNF334 Polyclonal specifically detects ZNF334 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| ZNF334 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| DKFZp586G1122, Hzf, HZFzinc finger protein 385, RZFHematopoietic zinc finger protein, ZFP385, zinc finger protein 385A, ZNF385Retinal zinc finger protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55713 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZNF334 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: HTGEKHGVFNKCGRISIVKSNCSQCKRMNTKENLYECSEHGHAVSKNSHLIVHQ | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto