missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF331 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566560
This item is not returnable.
View return policy
Description
Immunogen sequence: NIHEIRASKRN SDRRSKSLGR NWICEGTLER PQRSRGRYVN QMIINYVKRP ATREGTPPRT HQRHHKENSF Highest antigen sequence identity to the following orthologs - mouse 31%, rat 31%.
ZNF331 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 12 C2H2-type zinc fingers and 1 KRAB domain. ZNF331 may be involved in transcriptional regulation. It may play a role in spermatogenesis. Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form one family of zinc finger proteins.
Specifications
| ZNF331 | |
| Polyclonal | |
| Unconjugated | |
| ZNF331 | |
| C2H2-like zinc finger protein rearranged in thyroid adenomas; DKFZp686L0787; KRAB zinc finger protein; RITA; Zinc finger protein 331; zinc finger protein 361; Zinc finger protein 463; ZNF331; ZNF361; ZNF463 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55422 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQX6 | |
| ZNF331 | |
| Recombinant Human ZNF331. Recombinant protein control fragment (Product #RP-105256). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction