missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF329 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
ZNF329 Polyclonal antibody specifically detects ZNF329 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | ZNF329 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | zinc finger protein 394, zinc finger protein 99, Zinc finger protein with KRAB and SCAN domains 14, ZKSCAN14FLJ12298 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: REKPYKYPESVKSFNHFTSLGHQKIMKRGKKSYEGKNFENIFTLSSSLNENQRNLPGEKQY |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?