missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF329 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560693
This item is not returnable.
View return policy
Description
Immunogen sequence: EHLIASSDLP PSQRVLATNG FHAPDSNVSG LDCDPALPSY PKSYADKRTG DSDACGKGFN HSMEVIHGR Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 48%.
Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Krueppel C2H2-type zinc finger protein family, ZNF329 (zinc finger protein 329) is a 541 amino acid nuclear protein that contains twelve C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
Specifications
| ZNF329 | |
| Polyclonal | |
| Unconjugated | |
| ZNF329 | |
| 2810439M05Rik; 4632409L22Rik; RGD1565029; Zfp329; Zinc finger protein 329; Znf329 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UD4 | |
| ZNF329 | |
| Recombinant protein corresponding to Human ZNF329. Recombinant protein control fragment (Product #RP-93629). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction