missing translation for 'onlineSavingsMsg'
Learn More

ZNF329 Antibody (OTI6B12) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30687686 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687686 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687686 Supplier Novus Biologicals Supplier No. NBP344975AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF329 Monoclonal antibody specifically detects ZNF329 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF329
Applications Western Blot
Classification Monoclonal
Clone OTI6B12
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias zinc finger protein 394, zinc finger protein 99, Zinc finger protein with KRAB and SCAN domains 14, ZKSCAN14FLJ12298
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-203 of human ZNF329 (NP_078896) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79673
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.