missing translation for 'onlineSavingsMsg'
Learn More

ZNF329 Antibody (OTI2B11), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen ZNF329
Clone OTI2B11
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306512
View Documents
Novus Biologicals
NBP3-44986-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306559
View Documents
Novus Biologicals
NBP3-44986-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF329 Monoclonal antibody specifically detects ZNF329 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications

Specifications

ZNF329
Western Blot 1:2000, Immunohistochemistry 1:150
Monoclonal
Purified
RUO
PBS (pH 7.3) 1% BSA, 50% glycerol
79673
IgG1
>95%
Antibody
OTI2B11
Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human
zinc finger protein 394, zinc finger protein 99, Zinc finger protein with KRAB and SCAN domains 14, ZKSCAN14FLJ12298
Human recombinant protein fragment corresponding to amino acids 1-203 of human ZNF329 (NP_078896) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.